Product Image
Thermo Scientific™
Remel™ EE Broth Mossel
Thermo Scientific™ Remel EE Broth Mossel is used for enrichment and selective isolation of Enterobacteriaceae.
Have Questions?
Catalog NumberFormat
R112280PB - 100mL
Catalog number R112280
Price (USD)
-
Format:
PB - 100mL
Thermo Scientific™ Remel EE Broth Mossel is used for enrichment and selective isolation of Enterobacteriaceae.
Not intended for IVD use.
Specifications
DescriptionEE Broth Mossel
FormatPB - 100mL
Quantity10/Pk.
Product TypeMicrobiology Media
Unit SizeEach