Product Image
Thermo Scientific™
Remel™ TT Broth Base
Thermo Scientific™ Remel TT Broth Base (Hajna) Basal medium is used for selective enrichment and isolation of Salmonella.
Have Questions?
Catalog NumberQuantity
R1123704 x 1000 mL
Catalog number R112370
Price (USD)
-
Quantity:
4 x 1000 mL
Thermo Scientific™ Remel TT Broth Base (Hajna) Basal medium is used for selective enrichment and isolation of Salmonella.
Requires Iodine Solution, REF R114352. Not intended for IVD use.
Specifications
Certifications/ComplianceIndustrial Reference: USDA.
DescriptionTryptose Phosphate Broth
FormatBorosilicate Bottle
Quantity4 x 1000 mL
Product TypeMicrobiology Media
Unit SizeEach