Product Image
Thermo Scientific™

R2A Agar

Enumerate and cultivate heterotrophic bacteria from potable water using R2A Agar.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
R11254310 x 500 mLBB - 500mL
R0172210/Pk.Monoplate
R0172310/Pk.15mm x 60mm Plate
R110722100/Pk.Monoplate
R11254210 x 200 mLBB - 200mL
Catalog number R112543
Price (USD)
-
Quantity:
10 x 500 mL
Format:
BB - 500mL

Enumerate and cultivate heterotrophic bacteria from potable water using Thermo Scientific™ Remel™ R2A Agar. The growth of stressed and chlorine-tolerant bacteria recovered from treated waters is stimulated using a less nutritious medium, lower incubation temperatures, and longer incubation times. R2A Agar is recommended by the American Public Health Association (APHA) in Standard Methods for the Examination of Water and Wasterwater1. Information on water quality, such as heterotrophic plate counts, may be used to evaluate the efficiency of treatment processes.

Use R2A Agar for enumeration and cultivation of heterotrophic bacteria from potable water.

  • Recommended by the APHA in Standard Methods for the Examination of Water and Wasterwater1
  • Superior to plate count agar when using the pour plate, spread plate and the membrane filter techniques3.
  • Ready-to-use convenience of prepared media.

Reasoner and Geldreich of the U.S. Environmental Protection Agency developed R2A Agar for the isolation and recovery of aerobic and facultative heterotrophic bacteria from treated potable water samples2. R2A Agar was found to be superior to Plate Count Agar when using the pour plate, spread plate, or membrane filter techniques3. In this medium the low nutrients give higher counts in treated potable waters as compared to a highly nutritious medium because these bacteria tend to be slow-growing species and would quickly be suppressed by faster-growing species on a richer culture medium.

This medium contains peptone that supplies nitrogenous compounds and amino acids necessary for growth. Dextrose is added as a carbon source of energy. Yeast extract supplies B-complex vitamins and serves as a growth enhancer. Addition of soluble starch as a neutralizer and sodium pyruvate stimulates the growth of stressed bacteria. Buffering agents such as dipotassium phosphate and magnesium sulfate maintain osmotic equilibrium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

  1. Clesceri, L.S., A.E. Greenberg, and A.D. Eaton. 1998. Standard Methods for the Examination of Waste and Wastewater. 20th APHA. Washington, D.C.
  2. Reasoner, D.J. and E.E. Geldreich. 1979. Abstracts #N7. Abstracts of the 79th General Meeting of the American Society for Microbiology. ASM, Washington, D.C.
  3. Reasoner, D.J. and E.E. Geldreich. 1985. Appl. Environ. Microbiol. 49:1-7.
Specifications
Certifications/ComplianceIndustrial Reference: SMWW.
DescriptionR2A Agar
FormatBB - 500mL
Quantity10 x 500 mL
Product TypeAgar
Unit SizeEach