Double Bagged Bottled Sabouraud Dextrose Medium
Thermo Scientific™

Double Bagged Bottled Sabouraud Dextrose Medium

Perform confident sterility testing with double bagged bottled media for added security between environmental class changes.
Have Questions?
Catalog NumberFormat
R112553Double Bagged Borosilicate Bottle
Catalog number R112553
Price (USD)
-
Format:
Double Bagged Borosilicate Bottle
Perform confident sterility and microbial limits testing with Thermo Scientific™ Remel™ Double Bagged Bottled Sabouraud Dextrose Medium for testing according to USP, JP, and EU regulations and as a general purpose enrichment broth for cultivation and isolation of microorganisms. Double Bagged Sabouraud Dextrose Medium is ideal for microbiological testing that requires additional packaging for sterility protection between environmental class changes.
Add Remel Double Bagged Bottled Sabouraud Dextrose Medium to your sterility and microbial limits testing routine. Our range of validated solutions for the pharmaceutical microbiology laboratory support the production and quality assurance of sterile pharmaceuticals, vaccines, and biotechnology products.

Complete Solutions, Complete Confidence:
  • Secure packaging: double bagged prior to autoclaving to preserve integrity of sterility media
  • Regulatory compliant: USP, JP, and EU harmonized media  
  • Adaptable: a range of available fill volumes, media and bottle types to accommodate your sample sizes and performance testing needs
  • Quality guaranteed: Certificates of Analysis with every lot produced  
  • Easy-to-use: simply ‘peel and pop’ bags for quick access to the media bottle
  • Multi-purpose media: suitable for general quality testing
Customizable formats and media types are available. Please inquire.

Sabouraud Dextrose Medium is an acidic pH medium for the isolation of dermatophytes, other fungi, and yeasts.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use
For Laboratory Use
Specifications
Cap TypePolypropylene plastic cap w/Teflon
DescriptionDouble Bagged Bottled Sabouraud Dextrose Medium
FormatDouble Bagged Borosilicate Bottle
Quantity100 mL
Shelf Life365 Days
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 8°C