Search
Search
Cultivates pathogenic yeast and fungi from clinical specimens with Sabouraud Dextrose Broth 2%.
| Catalog Number | Quantity | Format |
|---|---|---|
| R112559 | 10 x 90 mL | WMB Lug - 90mL |
| R064410 | 100/Pk. | 15mm x 103mm Tube |
| R064412 | 20/Pk. | 15mm x 103mm Tube |
| R112554 | 10 x 100 mL | WMB Lug - 100mL |
| R112558 | 4 x 990 mL | Borosilicate Bottle - 990mL |
Cultivates pathogenic yeast and fungi from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Broth 2%. Sabouraud Dextrose Agar is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concentration of dextrose from 40 to 20g/L2. Sabouraud Dextrose Broth 2% is a modification of the original formulation, without agar.
Cultivate pathogenic yeast and fungi from clinical specimens with Sabouraud Dextrose Broth 2%.
This medium contains casein and meat peptones nitrogenous compounds and amino acids necessary for the growth of yeasts and fungi. Dextrose is an energy source which serves to support the growth of fungi. The low pH of the medium is favorable to the growth of fungi, especially dermatophytes, while also inhibiting contaminating bacteria in clinical specimens.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.