Search
Search
| Catalog Number | Quantity | Format | Packaging Type |
|---|---|---|---|
| R112732 | 10 x 500 mL | BB Septum - 500mL | 10/Pk. |
| R08943 | 100 x 15 mL | 20mm x 113mm Tube | 100/Pk. |
| R112728 | 10 x 90 mL | WMB Screw - 90mL | 10/Pk. |
| R112730 | 10 x 100 mL | WMB Lug - 100mL | 10/Pk. |
| R112731 | 10 x 100 mL | SB - 100mL | 10/Pk. |
| R112740 | 4 x 1000 mL | BB - 1000mL | 4/Pk. |
| R112745 | 10 x 100 mL | BB Septum - 100mL | 10/Pk. |
| R112758 | 10 x 25 mL | SB - 25mL | 10/Pk. |
| R112748 | 10 x 200 mL | WMB Lug - 200mL | 10/Pk. |
Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.