Double Bagged Bottled Tryptone Soya Broth (TSB)
Double Bagged Bottled Tryptone Soya Broth (TSB)
Double Bagged Bottled Tryptone Soya Broth (TSB)
Double Bagged Bottled Tryptone Soya Broth (TSB)
Thermo Scientific™

Double Bagged Bottled Tryptone Soya Broth (TSB)

Perform confident sterility testing with double bagged bottled media for added security between environmental class changes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormatCap Type
R112912Dbl Bagged Borosilicate BottlePP plastic cap w/ Teflon
R112996Dbl Bagged Wide Mouth BottleWhite Metal
R112751Double Bagged Serum BottleStopper, Crimp seal
Catalog number R112912
Price (USD)
-
Format:
Dbl Bagged Borosilicate Bottle
Cap Type:
PP plastic cap w/ Teflon
Perform confident sterility and microbial limits testing with Thermo Scientific™ Remel™ Double Bagged Bottled Tryptone Soya Broth (TSB) for testing according to USP, JP, and EU regulations and as a general purpose enrichment broth for cultivation and isolation of microorganisms. Double Bagged Bottled Tryptone Soya Broth (TSB) is ideal for microbiological testing that requires additional packaging for sterility protection between environmental class changes.
Add Remel Double Bagged Bottled Tryptone Soya Broth (TSB) to your sterility and microbial limits testing routine. Our range of validated solutions for the pharmaceutical microbiology laboratory support the production and quality assurance of sterile pharmaceuticals, vaccines, and biotechnology products.

Complete Solutions, Complete Confidence:
  • Secure packaging: double bagged prior to autoclaving to preserve integrity of sterility media
  • Regulatory compliant: USP, JP, and EU harmonized media
  • Adaptable: a range of available fill volumes, media and bottle types to accommodate your sample sizes and performance testing needs
  • Quality guaranteed: Certificates of Analysis with every lot produced
  • Easy-to-use: simply ‘peel and pop’ bags for quick access to the media bottle
  • Multi-purpose media: suitable for general quality testing
Customizable formats and media types are available. Please inquire.

Tryptone Soya Broth is a highly nutritious versatile medium which is recommended for general laboratory use. Due to the inclusion of both Tryptone and Soya Peptone, the medium will support a luxuriant growth of many fastidious organisms without the addition of serum.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use
For Laboratory Use
Specifications
Cap TypePP plastic cap w/ Teflon
DescriptionDouble Bagged Bottled Tryptone Soya Broth (TSB)
FormatDbl Bagged Borosilicate Bottle
Quantity100 mL
Shelf Life365 Days
Product TypeBroth
Unit SizeEach
Contents & Storage
2 to 25°C