Remel™ Butterfield's Phosphate Buffer
Actual product may vary
Thermo Scientific™

Remel™ Butterfield's Phosphate Buffer

Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatPackaging Type
R117061100 x 9 mLGlass Vial - 9mL100/Pk.
R11251910 x 100 mLPB -100mL10/Pk.
R1125201000 mLPB -1000mLEach
R11252610 x 90 mLPB -90mL10/Pk.
Catalog number R117061
Price (USD)
-
Quantity:
100 x 9 mL
Format:
Glass Vial - 9mL
Packaging Type:
100/Pk.

Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.

Not intended for IVD use.
Specifications
Certifications/ComplianceIndustrial Reference: BAM, USDA, USP.
DescriptionPhosphate Buffer, Butterfield's
FormatGlass Vial - 9mL
Packaging Type100/Pk.
Quantity100 x 9 mL
Product TypeBuffer Solution
Unit SizeEach