Search
| Catalog Number | Quantity | Format |
|---|---|---|
| R117880 | 20 x 9 mL | Glass vial-9mL |
| R112850 | 10 x 225 mL | BB - 225mL |
Thermo Scientific™ Remel UVM Modified Listeria Enrichment Broth is a selective enrichment for isolation of Listeria monocytogenes.
Ordering Information:
REF R117870 is polypropylene vial with screw cap.