Product Image
Thermo Scientific™

Remel™ Phosphate Buffer w/MgCl2 (pH 7.2) (Buffered Water) APHA Formulation

The Thermo Scientific™ Remel Phosphate Buffer with MgCl2 (pH 7.2) APHA Formulation is used as a diluent in microbial limit testing of food and water.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
R118610100 x 9 mL20mm x 113mm Tube
R0777010 x 90 mLBorosilicate Bottle, Septum
R11253110 x 20 mLSerum Bottle, Crimp
R11253010 x 20 mLSerum Bottle, Open Crimp
Catalog number R118610
Price (USD)
-
Quantity:
100 x 9 mL
Format:
20mm x 113mm Tube

Thermo Scientific™ Remel Phosphate Buffer with MgCl2 (pH 7.2) (Buffered Water) APHA Formulation is used as a diluent in microbial limit testing of food and water.

Ordering Information:

REF R112530 has open crimp seal. REF R112531 has complete crimp seal.

Specifications
Certifications/ComplianceIndustrial Reference: AOAC, APHA.
DescriptionPhosphate Buffer w/MgCl
Format20mm x 113mm Tube
Quantity100 x 9 mL
Product TypeBuffer Solution
Unit SizeEach