Product Image
Thermo Scientific™

Remel™ 2X Asparagine Broth

Use for the differentiation of nonfermentative gram-negative bacteria, especially Pseudomonas aeruginosa, in tube technique for recreational waters.
Have Questions?
Catalog NumberQuantity
R11900220 x 10 mL
Catalog number R119002
Price (USD)
-
Quantity:
20 x 10 mL

Thermo Scientific™ Remel 2X Asparagine Broth is for the differentiation of nonfermentative gram-negative bacteria, especially Pseudomonas aeruginosa, in the multiple tube technique for recreational waters.

Not for IVD use.
Specifications
Certifications/ComplianceIndustrial Reference: SMWW.
DescriptionAsparagine Broth, 2X
Format20mm x 150mm Tube
Quantity20 x 10 mL
Product TypeMicrobiology Media
Unit SizeEach