Resuspension Solution for GeneJET Plasmid Miniprep Kit
Thermo Scientific™
Resuspension Solution for GeneJET Plasmid Miniprep Kit
Thermo Scientific™ GeneJET Resuspension Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat No. K0502 and K0503) and may be purchased separately.
Have Questions?
Catalog NumberQuantity
R121370 mL
Catalog number R1213
Price (USD)
69.65
Online exclusive
72.75
Save 3.10
Each
Quantity:
70 mL
Price (USD)
69.65
Online exclusive
72.75
Save 3.10
Each
Thermo Scientific™ GeneJET Resuspension Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat No. K0502 and K0503) and may be purchased separately.
Highlights
  • The Resuspension Solution should be stored at 4°C and is stable for 6 months.
  • After the addition of RNase A, the Resuspension Solution is stable for 6 months when stored at 4°C.
Related Products
  • Neutralization Solution for GeneJET Plasmid Miniprep Kit (Cat. No. R1233)
  • Wash Solution for GeneJET Plasmid Miniprep Kit (R1243)
  • Binding Buffer for GeneJET Plasmid Miniprep Kit (R1331)
  • GeneJET NGS Cleanup Kit (K0851)
  • Binding Buffer for GeneJET NGS Cleanup Kit (R2811)
  • Prewash Buffer for GeneJET NGS Cleanup Kit (R2821)
  • Wash Buffer for GeneJET NGS Cleanup Kit (R2831)
  • GeneJET Plasmid Miniprep Kit (K0502)
  • RNase A (R1253)
  • Elution Buffer for GeneJET Plasmid Miniprep Kit (R1263)
  • Lysis Solution for GeneJET Plasmid Miniprep Kit (R1223)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSuspension Buffer
Product LineGeneJET
Product TypeResuspension Solution
Quantity70 mL
FormatKit
Unit SizeEach