An aerobic, facultative and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.
Have Questions?
Catalog Number
Quantity
R124101
10/Pk.
Catalog number R124101
Price (USD)
-
Quantity:
10/Pk.
Thermo Scientific™ Remel A.C.T. II is an aerobic, facultative, and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.
Fluids may be introduced through septum in cap
Sterile packaged in individual Tyvek™ envelope with single plastic-shaft rayon-tipped swab for clean environments
Do not freeze or overheat. Do not incubate prior to use.
For In Vitro Diagnostic Use.
Specifications
For Use With (Application)Healthcare
Quantity10/Pk.
Shaft MaterialPlastic
Storage RequirementsStore in original container at 15°C to 30°C until used.