MicroTest™ Flocked Swab Kits
Actual product may vary
Thermo Scientific™

MicroTest™ Flocked Swab Kits

Maintain organism viability with kits including quality room-temperature transport media, one traditional-tipped nylon flocked swab and/or one micro-tipped nylon flocked swab.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTip TypeProduct TypeIncludesFill Volume
R12542MicrotipFlocked SwabFlock Swab, Nylon MicroTip
R12565TraditionalFlocked Swab KitM4RT Kit w/Traditional-Tipped Flocked Swab3 mL
R12567Traditional and MicrotippedUniversal KitM4RT Universal Kit w/Micro-Tipped and Traditional-Tipped Flocked Swab3 mL
R12561MicrotipFlocked Swab KitM6 Kit w/Micro-Tipped Flocked Swab1.5 mL
R12569MicrotipFlocked Swab KitM6 Kit w/Micro-Tipped Flocked Swab3 mL
R12560TraditionalFlocked Swab KitM6 Kit w/Traditional-Tipped Flocked Swab1.5 mL
R12568TraditionalFlocked Swab KitM6 Kit w/Traditional-Tipped Flocked Swab3 mL
R12562Traditional and MicrotippedFlocked Swab KitM6 Universal Kit w/Micro-Tipped and Traditional-Tipped Flocked Swab1.5 mL
R12563Traditional and MicrotippedUniversal KitNylon MicroTip3 mL
Catalog number R12542
Price (USD)
-
Tip Type:
Microtip
Product Type:
Flocked Swab
Includes:
Flock Swab, Nylon MicroTip

Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses chlamydiae, mycoplasma and ureaplasma with quality media, proven collection and transport tools, and safe, reliable packaging with Thermo Scientific™ MicroTest™ Flocked Swab Kits.

Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.

MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.

MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

Microtest Flocked Swab Kits come with 1.5 mL or 3 mL of liquid medium in various tube types, with or without glass beads, individually wrapped swabs, in either a peel pouch format or zip-seal bag. Please refer to the specific product listing for more information about the kit contents.

Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.

Trusted reliable results

  • Stringent quality control, including cellular toxicity on each lot number
  • Patented formulations with stabilizing proteins, buffers, and specific antibiotics

Compatibility and viability

  • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
  • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing

Convenient, accommodating format

  • 1.5 mL and 3.0 mL fill volumes allow multiple tests from one specimen
  • Individual transport media tubes or complete kits offer flexibility and versatility

Safe, sturdy packaging

  • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
For Use With (Application)Viruses: Chlamydiae, Ureaplasmas, and Mycoplasmas
IncludesFlock Swab, Nylon MicroTip
Quantity100/Pk.
Storage Requirements5°C to 25°C
Tip TypeMicrotip
Product TypeFlocked Swab
Unit SizeEach