Do you offer the binding buffer included in the GeneJET PCR Purification Kit as a stand-alone product?
Yes, we offer Binding Buffer for GeneJET Gel Extraction Kit as a stand-alone product: Cat. No. R1331.
Search
View additional product information for Binding Buffer for GeneJET Gel Extraction Kit - FAQs (R1331)
1 product FAQs found