Product Image
Thermo Scientific™

Wash Buffer for GeneJET Gel Extraction Kit

Thermo Scientific™ GeneJET Wash Buffer is a component of the GeneJET Gel Extraction Kit and may be purchased separately. Provided in 45 mL aliquots.
Have Questions?
Catalog NumberQuantity
R134145 mL
Catalog number R1341
Price (USD)
72.75
Each
Quantity:
45 mL
Price (USD)
72.75
Each

Thermo Scientific™ GeneJET Wash Buffer is a component of the GeneJET Gel Extraction Kit (Cat No. K0691, K0692) and may be purchased separately. Provided in 45 mL aliquots.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chemical Name or MaterialWash Buffer
For Use With (Equipment)GeneJET Gel Extraction Kit
Product LineGeneJET
Quantity45 mL
Solution TypeBuffer
Unit SizeEach