Product Image
Thermo Scientific™
Remel™ A3B Broth Liquid
The Thermo Scientific™ Remel A3B Broth Liquid is supplied as a liquid, for transport of Ureaplasma.
Have Questions?
Catalog NumberQuantity
R203036 x 1.8 mL
Catalog number R20303
Price (USD)
-
Quantity:
6 x 1.8 mL
The Thermo Scientific™ Remel A3B Broth Liquid is supplied as a liquid, for transport of Ureaplasma.
Specifications
DescriptionA3B Broth
FormatVial - 1.8mL
Quantity6 x 1.8 mL
Product TypeMicrobiology Media
Unit SizeEach