Product Image
Thermo Scientific™
Arginine Broth
Thermo Scientific™ Remel Arginine Broth (Lyophilized) is used for cultivation and presumptive identification of Mycoplasma hominis.
Have Questions?
Número de catálogoCantidad
R203586 x 1,8 mL
Número de catálogo R20358
Precio (MXN)
-
Cantidad:
6 x 1,8 mL
Thermo Scientific™ Remel Arginine Broth (Lyophilized) is used for cultivation and presumptive identification of Mycoplasma hominis.
Especificaciones
DescripciónArginine Broth; lyophilized
FormatoVial
Cantidad6 x 1,8 mL
Tipo de productoMicrobiology Media
Unit SizeEach