Rapid Urea Broth (Lyophilized)
Thermo Scientific™
Rapid Urea Broth (Lyophilized)
Differentiate microorganisms on the basis of urease production using Rapid Urea Broth (Lyophilized).
Have Questions?
Catalog NumberQuantity
R203883 mL
Catalog number R20388
Price (USD)
-
Quantity:
3 mL
Rapidly differentiate microorganisms on the basis of urease production with Thermo Scientific™ Remel™ Rapid Urea Broth (Lyophilized). In 1941 Rustigian and Stuart developed a medium for detection of urease activity by Proteus spp.1 They further observed that other urea-positive genera within Enterobacteriaceae could be detected in a medium with reduced buffering capacity2. Rapid Urea Broth provides such a medium and allows rapid urease detection in enteric gram-negative bacilli other than Proteus spp. This medium is recommended by the American Public Health Association3.
The detection of urease activity is routinely used in clinical microbiology to aid in identification of gram-negative bacteria. Rapid Urea Broth (Lyophilized) is a medium recommended for use in rapid differentiation of microorganisms on the basis of urease production. Microorganisms that hydrolyze urea by means of urease enzyme produce ammonia. This alkaline end product causes the pH indicator, phenol red, to change color from yellow-orange to pink-red.
A balance of monopotassium phosphate and disodium phosphate comprise the buffering system to produce a rapid change in pH that changes phenol red indicator accordingly.
  • Easy to use – Lyophilized medium. Reconstitute with 3mL demineralized water or neutral saline (pH 7.0 ± 0.2).
  • Easy to interpret – A pink red color development indicates a positive test. No color change indicates a negative test.
  • Recommended by the American Public Health Association (APHA)3.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Rustigian, R. and C. Stuart. 1941. Proc. Soc. Exp. Biol. Med. 47:108-112.
  2. Rustigian, R. and C. Stuart. 1945. J. Bacteriol. 49:419-444.
  3. Food and Drug Administration. 2000. Bacteriological Analytical Manual Online. Chapter 5, Updated Dec. 2007. Authors: Andrews, W.H. and T.S. Hammack. AOAC International, Gaithersburg, MD.
Specifications
DescriptionUrea Broth, Rapid
FormatVial
Quantity3 mL
Product TypeMicrobiology Media
Unit SizeEach