Product Image
Thermo Scientific™
Penicillin Disk
Confirm Gram stain reactions and presumptively identify anaerobic, gram-negative bacilli with this ready-to-use Penicillin Disk.
Have Questions?
KatalognummerMenge
R2113625 Disketten/Pk.
Katalognummer R21136
Preis (EUR)
-
Menge:
25 Disketten/Pk.
Confirm Gram stain reaction of anaerobes and presumptively identify anaerobic, gram negative bacilli with Thermo Scientific™ Remel™ Penicillin Disk. Proposed in 1971 by Sutter and Finegold1 who recommended a method to identify anaerobic, gram-negative bacilli based on the difference in response to an antibiotic. Accordingly, organisms can be grouped based on their characteristic inhibitory patterns with erythromycin, rifampin, colistin, penicillin, kanamycin and vacomycin2,3.
Presumptively identify anaerobic bacteria with Penicillin Disk3,4.
  • Ready to use – Vancomycin-impregnated disk
  • Easy to interpret – With visual inspection
Penicillin G Disk contains 2 U penicillin.
Disks impregnated with erythromycin, rifampin, colistin, penicillin, kanamycin and vancomycin can be used to identify anaerobic, gram negative bacilli based on the zone of inhibition they exhibit. Colistin and vancomycin confirm the gram stain reaction, which is helpful with the clostridia that stain gram-negative. Penicillin, rifampin and kanamycin separate Bacteroides and Fusobacterium.
General References:
  1. Sutter, V.L. and S.M. Finegold. 1971. Appl. Microbiol. 21:13-20.
  2. Leigh, D.A. and K. Simmons. 1977. J.Clin. Pathol. 30:991-992.
  3. Forbes, B.A., D.F. Sahm, and A.S. Weissfeld. 2002. Bailey and Scott's Diagnostic Microbiology. 11th ed. Mosby, St Louis, MO.
  4. Summanen, P., E.J. Baron, E.M. Citron, C.A. Strong, H.M. Wexler, and S.M. Finegold, 1993. Wadsworth Anaerobic Bacteriology Manual. 5th ed. Star Publ. Co. Belmont, CA.
Specifications
BeschreibungPenicillin Disk
Menge25 Disketten/Pk.
Unit SizeEach
Inhalt und Lagerung
2 °C bis 8 °C