Remel™ 12.5% Gelatin Reagent
Thermo Scientific™
Remel™ 12.5% Gelatin Reagent
The Thermo Scientific™ Remel 12.5% Gelatin Reagent, Acidified Mercuric Chloride is for detection of gelatinase activity of microorganisms.
Have Questions?
Catalog NumberQuantity
R2122625 mL/Bottle
Catalog number R21226
Price (USD)
-
Quantity:
25 mL/Bottle
The Thermo Scientific™ Remel 12.5% Gelatin Reagent, Acidified Mercuric Chloride is for detection of gelatinase activity of microorganisms.
Specifications
DescriptionGelatin Reagent, 12.5% (Acidified Mercuric Chloride); Volume: 25mL/bottle
FormatBottle
Quantity25 mL/Bottle
Unit SizeEach
Contents & Storage
Store at Room Temperature