Product Image
Thermo Scientific™

Remel™ Polysorbate 80, 10%

Differentiate and identify catalase producing Mycobacterium spp. using Polysorbate 80, 10% reagent.
Have Questions?
Número de catálogoCantidad
R2127510 mL
Número de catálogo R21275
Precio (MXN)
-
Cantidad:
10 mL
Differentiate and identify Mycobacterium spp. with the catalase test using Thermo Scientific™ Remel™ Polysorbate 80, 10% reagent. Most strains of mycobacteria produce catalase1. The catalase test is a semiqualitative test which was developed to help in differentiation of organisms included in Mycobacterium tuberculosis complex (drug-resistant strains) from other Mycobacterium spp2 (non-drug-resistant strains)3,4.
Polysorbate 80 helps in dispersing the hydrophobic, tightly clumped mycobacterial cells from large aggregates to individual bacilli, thus maximizing the detection of catalase. 10% Polysorbate 80 permits more rapid entry of compounds, such as 30% hydrogen peroxide into the mycobacterial cell by lowering the surface tension in a medium.Ready-to-use: Liquid reagentEasy to interpret:Strong Positive Test: A column of bubbles > 45mmWeak Positive Test: A column of bubbles < 45mmNegative Test: No bubblesThis reagent contains 0.2 g/1000mL of Polysorbate 80 (CAS 9005-65-6) supplied in 10mL bottles.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

General References:

  1. Isenberg, H.D. 2004. Clinical Microbiology Procedures Handbook. 2nd ASM Press, Washington, D.C.
  2. Wayne, L.G. and J.R. Doubek. 1968. Appl. Microbiol. 16:925-931.
  3. Kent, P.T. and G.P. Kubica. Public Health Mycobacteriology, A Guide for the Level III Laboratory. 1985. U. S. Dept. of H.H.S., CDC, Atlanta, GA.
  4. Forbes, B.A., D.F. Sahm, and A.S. Weissfeld. 2007. Bailey and Scott's Diagnostic Microbiology. 12th Mosby Elsevier, St. Lois, MO.
Especificaciones
DescripciónPolysorbate 80, 10%
FormatoVial
Cantidad10 mL
Unit SizeEach
Contenido y almacenamiento
De 2 °C a 8 °C