Remel™ Butterfield's 99mL Phosphate Buffer
Thermo Scientific™

Remel™ Butterfield's 99mL Phosphate Buffer

Formulation is used in food, dairy and pharmaceutical testing as referenced in AOAC, APHA, FDA, USDA, and USP test methods.
Have Questions?
Catalog NumberQuantity
R2370172 x 99 mL
Catalog number R23701
Price (USD)
-
Quantity:
72 x 99 mL

Thermo Scientific™ Remel Butterfield's 99mL Phosphate Buffer (pH 7.2 stock solution) formulation is used in food, dairy and pharmaceutical testing as referenced in AOAC, APHA, FDA, USDA and USP test methods.

Specifications
DescriptionPhosphate Buffer, Butterfield's 99mL
FormatWhite Cap - 99mL/Bottle
Packaging Type72/Case
Quantity72 x 99 mL
Product TypeBuffer Solution
Unit SizeEach