Thermo Scientific™ Remel Phosphate Buffer w/MgCl 90mL (pH 7.2 stock solution) formulation is used in water and wastewater testing as referenced in APHA and EPA test methods.
Have Questions?
Catalog Number
Quantity
R23702
72 x 90 mL
Catalog number R23702
Price (USD)
-
Quantity:
72 x 90 mL
Thermo Scientific™ Remel Phosphate Buffer w/MgCl 90mL (pH 7.2 stock solution) formulation is used in water and wastewater testing as referenced in APHA and EPA test methods.
Dilution Bottles are leakproof, wide mouth PET plastic with tear away sterility strip and easy, one-hand opening hinged lid
Gamma irradiated
Cap color-coded (gold) for easy and accurate visual identification