Thermo Scientific™ Remel Phosphate Buffer w/MgCl 99mL (pH 7.2) formulation is used in water and wastewater testing as referenced in APHA and EPA test methods.
Have Questions?
Catalog Number
Quantity
R23703
72 x 99 mL
Catalog number R23703
Price (USD)
-
Quantity:
72 x 99 mL
Thermo Scientific™ Remel Phosphate Buffer w/MgCl 99mL (pH 7.2 stock solution) formulation is used in water and wastewater testing as referenced in APHA and EPA test methods.
pH 7.2 stock solution
Formulation is used in water and wastewater testing as referenced in APHA and EPA test methods