Search
| Catalog Number | Quantity |
|---|---|
| R244125 | 25 Tests/Kit |
Remel ImmunoDOT Torch Panel is an EIA membrane test determining the presence of antibodies to Toxoplasma gondii, rubella virus, cytomegalovirus, and herpes simplex virus in serum and whole blood.