Xpect™ Giardia Test
Xpect™ Giardia Test
Xpect™ Giardia Test
Xpect™ Giardia Test
Thermo Scientific™
Xpect™ Giardia Test
Rapidly detect Giardia-specific antigen from fresh, frozen, or preserved human fecal specimens.
Have Questions?
Catalog NumberQuantity
R245002020 Tests/Kit
Catalog number R2450020
Price (CLP)
-
Quantity:
20 Tests/Kit
Optimize your workflow by rapidly detecting Giardia-specific antigen from fresh, frozen, or preserved human fecal specimens using Thermo Scientific™ Xpect™ Giardia Test. Report accurate results in 15 minutes during all stages of infection—all from a single specimen.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Giardia-specific antigen. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple walk-away procedure.
  • Clearly visible, easy-to-read results in just 15 minutes
  • Less than one minute hands-on time
  • 95.8 to 97.9% sensitivity
  • Single and combination immunoassays available
  • No specimen dilution required when using the recommended Formalin, SAF, or modified Cary-Blair w/Indicator transport media
Xpect Giardia uses antibody-coated color particles to bind with target antigen in the diluted sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionXpect Giardia Test
IncludesTest devices, conjugate (2.5mL), specimen dilution buffer (2.5mL), procedure card, disposable transfer pipettes and dilution tubes
Quantity20 Tests/Kit
Sample TypeFecal
TypeGiardia Testing Kit
Detectable AnalytesGiardia
Unit SizeEach
Contents & Storage
2°C to 8°C