Search
Detect and identify acid fast organisms in fecal material using DMSO Modified Acid Fast Decolorizer with DMSO Modified Acid-Fast Stain Kit.
| Catalog Number | Quantity |
|---|---|
| R2463418 | 250 mL |
Use Thermo Scientific™ Remel™ DMSO Modified Acid Fast Decolorizer with DMSO Stain Kit (R246303) to detect and identify acid fast organisms (i.e., Cryptosporidium, Isospora, and Cyclospora spp.) in fecal material. It is a modification of Kinyoun method and may be also used for detecting acid-fast bacilli (Mycobacterium spp.) in sputum, urine, gastric lavage, other body fluids and tissues.
Cryptosporidium is a significant enteric pathogen that causes acute diarrhea in immunodeficient patients and transient diarrhea in immunocompetent individuals, particularly youngsters1-5. The need to facilitate appropriate treatment for infected patients has led to the development of rapid methods for detection of these organisms.
The DMSO Modified acid-fast stain is a rapid and permanent differential staining procedure for detection and identification of acid-fast organisms. DMSO method is reliable and consistent in detecting cryptosporidiosis. This method also detects other pathogens such as Cyclospora and Isospora6.
Dimethylsulfoxide (DMSO) added to carbolfuchsin enhances penetration of acid-fast organisms by the stain and eliminates the need for heat or steam in the staining process. The stain is removed from background material by the decolorizing solution.
This reagent contains 10% Sulfuric acid supplied in 250mL bottles.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.