Product Image
Thermo Scientific™
Remel™ Giemsa Plus Stain Kit
Thermo Scientific™ Remel Giemsa Plus Stain Kit is a modified Wright stain for detection and identification of blood and tissue parasites.
Have Questions?
Catalog NumberQuantity
R2464033/Pk.
Catalog number R246403
Price (USD)
-
Quantity:
3/Pk.
Thermo Scientific™ Remel Giemsa Plus Stain Kit is a modified Wright stain for detection and identification of blood and tissue parasites.
Includes: 1 bottle Giemsa Plus Fixative; 1 bottle Giemsa Plus Reagent A; 1 bottle Giemsa Plus Reagent B.
Specifications
Quantity3/Pk.
Product TypeGiemsa Plus Stain Kit
Volume (Metric)250 mL/Bottle
Unit SizeEach