Product Image
Thermo Scientific™
Remel™ ImmunoWELL™ Borrelia
Use for qualitative and semi-quantitative detection of Borrelia burgdorferi IgG, IgM and IgA antibodies in serum for the diagnosis of Lyme disease.
Have Questions?
Catalog NumberQuantity
R247043796 Wells
Catalog number R2470437
Price (USD)
-
Quantity:
96 Wells
Remel ImmunoWELL Borrelia is an EIA microwell test used for qualitative and semi-quantitative detection of Borrelia burgdorferi IgG, IgM and IgA antibodies in serum for the diagnosis of Lyme disease.
Manufacturer: GenBio
Specifications
DescriptionImmunoWELL™ Borrelia
Packaging Type1 Kit
Quantity96 Wells
TypeBorrelia Testing Kit
Detectable AnalytesB. burgdorferi IgA, B. burgdorferi IgG, B. burgdorferi IgM
Unit SizeEach