Remel™ Oxy-Swab™
Remel™ Oxy-Swab™
Remel™ Oxy-Swab™
Remel™ Oxy-Swab™
Thermo Scientific™
Remel™ Oxy-Swab™
For detection of cytochrome oxidase in oxidase-producing microorganisms, such as Neisseria spp. and Pseudomonas spp.
Have Questions?
Catalog NumberQuantity
R2493484 x 25 Swabs
Catalog number R249348
Price (USD)
-
Request A Quote
Quantity:
4 x 25 Swabs
Thermo Scientific™ Remel Oxy-Swab is used for detection of cytochrome oxidase in oxidase-producing microorganisms, such as Neisseria spp. and Pseudomonas spp.
Ordering Alerts:
Packaged in resealable foil envelopes. 25 Swabs/Envelope.
Specifications
DescriptionDetects cytochrome oxidase in oxidase-producing microorganisms, such as Neisseria spp. and Pseudomonas spp.
Packaging Type4 Env/Pk.
Product LineRemel, Oxy-Swab
Quantity4 x 25 Swabs
Detectable AnalytesCytochrome Oxidase
Unit SizeEach
Contents & Storage
2°C to 8°C