Zeocin™ Selection Reagent, 50 mL - FAQs
Zeocin™ Selection Reagent, 50 mL - FAQs
View additional product information for Zeocin™ Selection Reagent - FAQs (R25005, R25001)
11 product FAQs found
Which of your antibiotics (Geneticin, Zeocin, Hygromycin B, Blasticidin, and Puromycin) can be used together for stable selection in mammalian cells?
All of our antibiotics (Geneticin, Zeocin, Hygromycin B, Blasticidin, and Puromycin) can be used together for making multiple stable cell lines. However, kill curves will need to be performed for each combination of antibiotics since sensitivity to a given antibiotic tends to increase when combined with other antibiotics.
I received Zeocin and it looks blue. Is this normal?
Zeocin is a formulation of phleomycin D1, a basic, water-soluble, copper-chelated glycopeptide isolated from Streptomyces verticillus. The presence of copper gives the solution its blue color. This copper-chelated form is inactive. When the antibiotic enters the cell, the copper cation is reduced from Cu2+ to Cu1+ and removed by sulfhydryl compounds in the cell. Upon removal of the copper, Zeocin is activated and will bind DNA and cleave it, causing cell death.
What is the molecular weight of Zeocin?
MW=1,535. Molecular formula: C60H89N21O21S3.
What are the recommended concentrations of antibiotics to use for selection in prokaryotes and eukaryotes?
For best results, optimal concentrations for selection should be determined empirically in each unique experiment through dose response curves. However, to get a general idea of concentrations that have worked for individual cell types, please click on the following url: http://www.thermofisher.com/us/en/home/life-science/cell-culture/transfection/selection.html or type in “Selection Antibiotics” into our main search on www.thermofisher.com.
What safety precautions should be taken when autoclaving Zeocin-containing media and plates before disposal? Are there any long-term effects for people who are exposed to Zeocin on a daily basis?
Zeocin, in media or other liquid form, is a health hazard when it comes in contact with your skin or upon ingestion. When it is in the powder solid form, it can become a respiratory hazard as well as contact and ingestion hazard. If the liquid or media is placed in an autoclavable bag, it can be autoclaved. The elevated temperatures within the autoclave will inactivate Zeocin. Sodium hypochloride (bleach) will also inactivate Zeocin. We highly recommend wearing the appropriate personal protective equipment (lab coat, gloves, and eye protection) when handling Zeocin.
In contrast to Geneticin (G418)-induced cell death, cells treated with Zeocin do not always detach and float when they die. Is this typical?
It is true that a percentage of non-resistant mammalian cells do not round-up from the plate upon Zeocin selection as would be seen with G418 or Hygromycin selection. However, one should see some very characteristic morphological changes occurring in those cells that are not resistant. These cells that stick to the culture dish typically display a vast increase in size. This could be best described as being similar to the effects of cytomegalovirus infecting permissive cells. The shape of these cells may also change; taking on an "alien" shape. On close examination of the non-resistant cells, the researcher should observe a distinct breakdown of both the nuclear and plasma membranes. Even though the "cells" are still attached to the plate, they should have the appearance of many holes in these membranes. Also, before the breakdown of the membranes, one can observe open areas in the cytoplasm of the cells that appear to be large, empty vesicles. Although not confirmed, this may be explained by a breakdown of the endoplasmic reticulum and Golgi apparatus, or other scaffolding proteins. Eventually, these "cells" will completely breakdown so that only "strings" of protein are left.
In contrast, Zeocin resistant cells should continue to divide at a regular interval to form distinct clumps of cells, or colonies. There should not be a distinct change in morphology, which can be compared to cells not under selection with Zeocin. It is these colonies of actively dividing cells that contain the resistance gene and are expressing it actively.
If there is concern about the dead cells sticking to the plate, one may do the following to eliminate them: Treat the plate for a couple of minutes with trypsin/versene. Both the healthy resistant cells and the dead cells will dislodge from the plate. The cells can then be replated (without Zeocin selection) and the healthy cells will attach again while the dead ones will not. After a couple of hours when the healthy cells have attached to the substrate again, Zeocin can be added back to the medium.
If there are no zeocin-containing YPD plates readily available, would it be possible to spread zeocin on top of YPD plates and still retain efficient selection of yeast?
Zeocin reagent can be spread on top of YPD plates for selection of yeast if necessary. There is a report that this works well when done with 10-15 3mm glass beads. However, it is recommended that some optimization is performed since top-spreading may dilute the antibiotics' effectiveness.
What is the mode of action on the following antibiotics: Blasticidin, Geneticin (G418), Hygromycin, and Zeocin?
Blasticidin: Nucleoside Inhibits protein synthesis in prokaryotic and eukaryotic cells by interfering with peptidyl transfer reaction of protein synthesis, causing early termination of translation.
Geneticin (G418): Aminoglycoside Blocks protein synthesis in mammalian cells by interfering with ribosomal function.
Hygromycin: Aminocyclitol Inhibits protein synthesis by disrupting translocation and promoting mistranslation.
Zeocin: Intercalates with DNA and cleaves it.
I received my shipment of Zeocin Selection Reagent (Cat. No. R25001, R25005), and the date written on the tube has passed already. Does this mean it is expired?
The date on the tube is the date of manufacture, not expiration. This is shown by the image of a building on the label, which is indicates a manufacturing facility.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How can I decontaminate my cultures?
When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.
1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.
The following is a suggested procedure for determining toxicity levels and decontaminating cultures:
1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What antibiotics do you offer to help control or eliminate cell culture contamination?
Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.