Rat IgG2a Isotype Control, Alexa Fluor™ 488 - FAQs
Rat IgG2a Isotype Control, Alexa Fluor™ 488 - FAQs
View additional product information for Rat IgG2a Isotype Control, Alexa Fluor™ 488 - FAQs (R2A20)
This product does not have any FAQs available.