You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Microbial Identification Test Kits
▸
Neisseria Testing
▸
Remel™ Agglutinating Sera, Neisseria meningitidis
Thermo Scientific™
Remel™ Agglutinating Sera, Neisseria meningitidis
Used for serological identification of
N. meningitidis
by rapid slide agglutination and CIE procedures.
Have Questions?
Change view
Catalog Number
Description
R30167501
Group B
R30166901
Group C
R30166801
Group A
R30167401
Group W135
4 Options
Catalog number
R30167501
Price (USD)
-
Request A Quote
Description:
Group B
Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
Specifications
Description
Group B
Organism Group
Group B
Quantity
Each
Target Organism Class
Neisseria Meningitidis
Type
Neisseria Meningitidis Test Kit
CE Marker
Yes
Detectable Analytes
N. meningitidis
Unit Size
Each