Product Image
Thermo Scientific™

Remel™ Agglutinating Sera, Salmonella O Polyvalent

Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
R30858101Polyvalent (group A - G)
R30858201Polyvalent (group A - S)
Catalog number R30858101
Price (USD)
-
Description:
Polyvalent (group A - G)
Remel Agglutinating Sera, Salmonella O is used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Control SetsPositive
DescriptionPolyvalent (group A - G)
MaterialPolyvalent
Organism GroupGroup A-G
Packaging Type2 mL/Vial
Quantity2 mL
Target Organism ClassSalmonella O
TypeAgglutinating Sera
Detectable AnalytesSalmonella O Antigen
Unit SizeEach