Wellcogen™ Strep B Rapid Latex Agglutination Test, Wellcogen Strep B 30 Pk
Thermo Scientific™
Wellcogen™ Strep B Rapid Latex Agglutination Test, Wellcogen Strep B 30 Pk
Rapidly detect antigens to Streptococcus Group B present in human cerebrospinal fluid (CSF), serum, urine or blood cultures by latex agglutination.
Have Questions?
Catalog NumberQuantity
R3085870130 Tests/Kit
Catalog number R30858701
Price (USD)
-
Quantity:
30 Tests/Kit
Wellcogen™ Strep B is a rapid latex test for use in the qualitative detection of antigen from group B streptococci, present in body fluids as a consequence of infection1 or in blood cultures. NOTE: Tests performed directly on clinical specimens are intended for screening purposes and should augment, not replace, culture procedures. Results must be used in conjunction with other data; e.g. symptoms, results of other tests, clinical impressions etc.
Use this rapid latex agglutination test in the qualitative detection of antigens from Group B streptococci, present in body fluids as a consequence of infection, or in blood cultures. The reagent consists of polystyrene latex particles which have been coated with antibodies specific to the Group B antigen.
These latex particles agglutinate in the presence of sufficient homologous antigen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionWellcogen Strep B Latex Agglutination Test
Organism GroupGroup B
Quantity30 Tests/Kit
TypeStreptococcus Test Kit
CE MarkerYes
Unit SizeEach
Contents & Storage
2°C to 8°C