Wellcogen™ Neisseria meningitidis B/E. coli K1 Rapid Latex Agglutination Test
Thermo Scientific™

Wellcogen™ Neisseria meningitidis B/E. coli K1 Rapid Latex Agglutination Test

Rapidly detect antigens to Neisseria meningitidis group B and E. coli K1 present in human body fluids, blood cultures, or on culture media by latex agglutination.
Have Questions?
Catalog NumberQuantity
R3085950230 Tests/Kit
Catalog number R30859502
Price (USD)
-
Quantity:
30 Tests/Kit
Wellcogen™ N. meningitidis B/E. coli K1 is a rapid latex test for use in the qualitative detection of antigen from Neisseria meningitidis (meningococcus) group B and Escherichia coli K1, present in body fluids as a consequence of infection. The latex can also be used for the detection of specific antigen in blood culture supernatants and for the identification of meningococcus group B or E. coli K1 cultures. Meningococcus group B antigen and E. coli K1 antigen are immunologically indistinguishable from each other. NOTE: Tests performed directly on clinical specimens are intended for screening purposes and should augment, not replace, culture procedures. Results must be used in conjunction with other data; eg symptoms, results of other tests, clinical impressions etc.
Use this rapid latex test in the qualitative detection of antigen from Neisseria meningitidis (meningococcus) group B and Escherichia coli K1 present in body fluids as a consequence of infection.

The latex can also be used for the detection of specific antigen in blood culture supernatants and for the identification of meningococcus group B or E. coli K1 cultures.

The reagent consists of polystyrene latex particles which have been coated with monoclonal antibodies specific to meningococcus group B antigen. These latex particles agglutinate in the presence of sufficient homologous antigen. N.B. The polysaccharide K1 antigen of E. coli shows structural and immunological identity with meningococcus group B antigen, and the reagent does not distinguish between them. Therefore, the reagent may be used to assist in the diagnosis of meningococcal meningitis and neonatal E. coli meningitis.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionWellcogen Neisseria meningitidis B/E. coli K1 Test Kit
Organism GroupGroup B
Quantity30 Tests/Kit
Target Organism ClassB/E. Coli
TypeNeisseria Meningitidis Test Kit
CE MarkerYes
Detectable AnalytesN. meningitidis (Group B) Antigen, E. coli K1 Antigen
Unit SizeEach
Contents & Storage
2°C to 8°C