Cary-Blair with Indicator Single Vial
Imagem ilustrativa. O produto pode variar
Cary-Blair with Indicator Single Vial
Cary-Blair with Indicator Single Vial
Cary-Blair with Indicator Single Vial
Thermo Scientific™
Cary-Blair with Indicator Single Vial
Collect, preserve, and recover enteric bacterial pathogens with a fecal transport that comes with industry color-coding and sporked caps and is stored at room temperature.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantityClosure Color
R3161012 Vials/Pk.Orange
R2161012 Vials/Pk.Green
R21925120 Vials/Pk.Green
R21617288 Vials/Pk.Green
R31925120 Vials/Pk.Orange
Número do catálogo R31610
Preço (BRL)
-
Quantity:
12 Vials/Pk.
Closure Color:
Orange
Collect, preserve, and recover enteric bacterial pathogens in stool specimens with Thermo Scientfic™ Cary-Blair with Indicator Single Vial, part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
Maintain the viability of enteric bacterial pathogens during transport and storage with a formulation that includes 1.16% agar and is ready for use without any special preparation. Save valuable storage space with a transport that is stored in its original container at room temperature.
  • Cary-Blair with Indicator is an isotonic and nonnutritive medium, which does not contain a fermentable carbohydrate.
  • Sodium thioglycollate is added to impede oxidation.
  • Disodium phosphate is incorporated in the medium to act as a buffering agent, which prevents overgrowth of Escherichia coli and Enterobacter species.
  • The relatively high pH minimizes the destruction of bacteria due to acid formation.
  • Phenol red, a pH indicator, is added to demonstrate acidic shifts that can affect the viability of some enteric pathogens.
Add convenience and safety to your fecal collection procedures with a broad line of transport products which have a long shelf life and are designed especially for enteric diagnostic testing.
Look to us for convenient kit packaging options, including bulk kits.
Multilingual patient collection instructions are included.
Choose from a wide range of collection and preservation products for fecal specimens, which is important to the successful growth, isolation and detection of microorganisms.
  • Cary Blair with Indicator
  • Formalin 10%
  • Formalin 5%
  • PVA Modified Fixative
  • SAF Fixative
  • Zinc PVA Fixative
  • Empty Vial
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Especificações
Closure ColorOrange
DescriptionCary-Blair with Indicator Single Vial (Orange Cap)
Fill Volume15 mL
Media TypeCary-Blair
PackagingSingle vial. Packaged in partitioned boxes.
Quantity12 Vials/Pk.
Product TypeFecal Transport Vial
Unit SizeEach