NucGreen™ Dead 488 ReadyProbes™ Reagent (SYTOX™ Green) - FAQs
NucGreen™ Dead 488 ReadyProbes™ Reagent (SYTOX™ Green) - FAQs
View additional product information for NucGreen™ Dead 488 ReadyProbes™ Reagent (SYTOX™ Green) - FAQs (R37109)
2 product FAQs found
Can I use the ReadyProbes reagents for flow cytometry?
This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.
What is the difference between NucBlue Live reagents stain and NucGreen Dead reagents stain?
The NucBlue Live reagent stains the nuclei of all cells, while NucGreen Dead reagent stains only the nuclei of dead cells.