NucRed™ Dead 647 ReadyProbes™ Reagent (TO-PRO-3 iodide) - FAQs
NucRed™ Dead 647 ReadyProbes™ Reagent (TO-PRO-3 iodide) - FAQs
View additional product information for NucRed™ Dead 647 ReadyProbes™ Reagent (TO-PRO-3 iodide) - FAQs (R37113)
1 product FAQs found
Can I use the ReadyProbes reagents for flow cytometry?
This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.