Annexin V Alexa Fluor™ 647 Ready Flow Conjugate, 120 reactions - FAQs
Annexin V Alexa Fluor™ 647 Ready Flow Conjugate, 120 reactions - FAQs
View additional product information for Annexin V Ready Flow Conjugates for Apoptosis Detection - FAQs (R37175, R37174, R37177, R37176)
3 product FAQs found
I would like to label only 100 µL of sample of the same cell density using a Ready Flow product. The manual notes 2 drops per 1 mL (1 X 10E6 cells/mL). May I scale down for smaller volumes?
Yes, you may scale up or down as needed, but we recommend keeping the cell density the same. There are 41 µL per drop, so 2 drops is 82 µL (for the 1 mL of sample). You need to only scale down for this 100 µL volume, which would be 8.2 µL.
What is the concentration of dye/reagent in the Ready Flow products?
The amount of dye or reagent in the Ready Flow products is proprietary. If you need to consider the exact amount of dye or reagent for your experiment, each of the Ready Flow reagents is available as a standalone product.
Can I use the ReadyProbes reagents for flow cytometry?
This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.