ReadyProbes™ In Situ Hybridization (ISH) Blocking Solution (5X) - FAQs
ReadyProbes™ In Situ Hybridization (ISH) Blocking Solution (5X) - FAQs
View additional product information for ReadyProbes™ In Situ Hybridization (ISH) Blocking Solution (5X) - FAQs (R37620)
2 product FAQs found
What is the volume for one drop from the ReadyProbes dropper bottles?
The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.
Can the ReadyProbes In Situ Hybridization (ISH) Blocking Solution (5X) be used with PrimeFlow and ViewRNA labeling?
We do not recommend using the ReadyProbes In Situ Hybridization (ISH) Blocking Solution (5X) with PrimeFlow and ViewRNA labeling as the PrimeFlow and ViewRNA kits utilize a specific blocking reagent optimized for these kits.