ReadyProbes™ Solvent-Resistant Permanent Marking Pen - FAQs
ReadyProbes™ Solvent-Resistant Permanent Marking Pen - FAQs
View additional product information for ReadyProbes™ Solvent-Resistant Permanent Marking Pen - FAQs (R37623)
1 product FAQs found
When would I use the ReadyProbes Solvent-Resistant Permanent Marking Pen versus a standard hydrophobic barrier pen?
We recommend using the ReadyProbes Solvent-Resistant Permanent Marking Pen if you will be subjecting your samples to any solutions containing alcohol or xylene.