ReadyProbes™ 2.5% Normal Horse Serum (1X) - FAQs
ReadyProbes™ 2.5% Normal Horse Serum (1X) - FAQs
View additional product information for ReadyProbes™ 2.5% Normal Horse Serum (1X) - FAQs (R37625)
4 product FAQs found
What is the volume for one drop from the ReadyProbes dropper bottles?
The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.
Can I use ReadyProbes 2.5% Normal Horse Serum (1X) as an antibody diluent for both primary and secondary antibodies?
Yes, ReadyProbes 2.5% Normal Horse Serum (1X) can be used as an antibody diluent for both primary and secondary antibodies, except for HRP-conjugated antibodies. The sodium azide added as a preservative is an inhibitor of horseradish peroxidase.
How many drops of ReadyProbes 2.5% Normal Horse Serum (1X) should I add to a sample?
Add a sufficient number of drops of ReadyProbes 2.5% Normal Horse Serum (1X) to completely cover the sample.
When would I use goat, horse, or chicken serum for blocking versus FBS or BSA?
For blocking, you should use a serum of the same or closely related species of the secondary antibody being used. If using a secondary antibody that was raised in goat or sheep, use goat serum (Cat. No. R37624 or 31873) for blocking; if using a secondary antibody raised in donkey or horse, use horse serum (Cat. No. R37625 or 31874) for blocking; if using a secondary antibody raised in chicken, use chicken serum (Cat. No. R37626) for blocking. FBS and BSA are alternatives when a specific species of serum is not available.