ReadyProbes™ Hydrophobic Barrier Pap Pen
ReadyProbes™ Hydrophobic Barrier Pap Pen
ReadyProbes™ Hydrophobic Barrier Pap Pen
ReadyProbes™ Hydrophobic Barrier Pap Pen
Thermo Scientific™

ReadyProbes™ Hydrophobic Barrier Pap Pen

The ReadyProbes Hydrophobic Barrier Pap Pen creates a water-repellent barrier when used on a microscope slide or coverslip. By lightlyRead more
Have Questions?
Catalog NumberQuantity
R37772 Pens (6 mL Each)
Catalog number R3777
Price (USD)
218.94
Special offer
Online exclusive
Ends: 31-Dec-2026
267.00
Save 48.06
Each
Quantity:
2 Pens (6 mL Each)
Price (USD)
218.94
Special offer
Online exclusive
Ends: 31-Dec-2026
267.00
Save 48.06
Each
The ReadyProbes Hydrophobic Barrier Pap Pen creates a water-repellent barrier when used on a microscope slide or coverslip. By lightly drawing a rectangle or circle around the tissue section or cells on the microscope slide, aqueous incubation reagents can be kept inside the circle or rectangle. Save precious reagents by incubating with less volume. The ReadyProbes Hydrophobic Barrier Pap Pen can also be used to keep reagents separate when more than one tissue section is mounted on a microscope slide.

The light blue barrier line is heat resistant and compatible with most immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) reagents, including detergent-containing reagents. It is insoluble in alcohol and acetone, but can be removed with clearing agent (e.g., xylene).

To use, dab the nib of the pen on a paper towel, to make sure the flow of the hydrophobic fluid is uninterrupted. Lightly trace a circle or rectangle around the tissue/cells 2-4 times, with a thick line, while keeping a few millimeters distance from the edge of the tissue/cells. Let the line dry for 20-30 minutes at room temperature. Repeat the process, if needed.

Specifications
Shelf Life6 months from receipt of product, or as stated on COA
Shipping ConditionRoom Temperature
For Use With (Application)Immunohistochemistry (IHC), Immunocytochemistry (ICC), Immunofluorescence (IF)
Product LineReadyProbes
Quantity2 Pens (6 mL Each)
Unit SizeEach
Contents & Storage
Store at room temperature, away from sunlight, or other heat sources.

Frequently asked questions (FAQs)

What is the volume for one drop from the ReadyProbes dropper bottles?

The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.