Product Image
Thermo Scientific™
Remel™ Gram Decolorizer
Thermo Scientific™ Remel Gram Decolorizer is a decolorizing reagent used in Gram stain procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R40054Each
R400555/Pk.
R40075Each
Catalog number R40054
Price (EUR)
-
Quantity:
Each
Thermo Scientific™ Remel Gram Decolorizer is a decolorizing reagent used in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
Specifications
Certifications/ComplianceCE Marked
QuantityEach
Product TypeDecolorizer
Volume (Metric)250 mL
Unit SizeEach