See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Biochemical Reagents
▸
Staining Reagents
▸
Stains and Dyes
▸
Remel™ Gram Safranin
Thermo Scientific™
Remel™ Gram Safranin
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
Have Questions?
Change view
Catalog Number
Volume (Metric)
R40058
250 mL
R40059
5 x 250 mL
R40079
3.785 L
3 Options
Catalog number
R40058
Price (USD)
-
Request A Quote
Volume (Metric):
250 mL
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
Selected Gram stains are CE marked
Specifications
Certifications/Compliance
CE Marked
Quantity
Each
Product Type
Safranin
Volume (Metric)
250 mL