This product requires proof of identity and completion of the DEA Iodine End Use Declaration before it can be purchased. Please complete this form if you have not previously done so.
Thermo Scientific™
Remel™ Gram Iodine
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
Have Questions?
Change view
Catalog Number
Quantity
R40077
Each
R40056
Each
R40057
5/Pk.
3 Options
Catalog number R40077
Price (USD)
-
Quantity:
Each
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
For qualitative procedures to differentiate gram-positive from gram-negative microorganisms