Product Image
Thermo Scientific™
Remel™ Gram Enhancer
Thermo Scientific™ Remel Gram Enhancer is used to reduce the red color of background debris to give better contrast to small gram-negative microorganisms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R40232Each
R40231250mL/Bottle
Catalog number R40232
Price (USD)
-
Quantity:
Each
Thermo Scientific™ Remel Gram Enhancer is used to reduce the red color of background debris to give better contrast to small gram-negative microorganisms.
Specifications
QuantityEach
Product TypeEnhancer
Volume (Metric)3.785 L
Unit SizeEach