This product requires proof of identity and completion of the DEA Iodine End Use Declaration before it can be purchased. Please complete this form if you have not previously done so.
Thermo Scientific™
Remel™ Enhanced Gram Iodine
Thermo Scientific™ enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
Have Questions?
Change view
Catalog Number
Quantity
R40235
Each
R40234
5/Pk.
2 Options
Catalog number R40235
Price (USD)
-
Quantity:
Each
Thermo Scientific™ Remel enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.